Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Screw Caps for 50 mL Centrifuge Tubes
GREENER_CHOICE

Catalog No. 07000524 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color: Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
07-000-524 Color: Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-524 Supplier Greiner Bio-One Supplier No. 346370
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with 50mL Centrifuge Tube

  • For use with 50mL Centrifuge Tube
  • Non-sterile
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Sterility Non-sterile
For Use With (Equipment) 50 mL Centrifuge Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.