Learn More
Market Lab Screw Caps for Graduated Tubes, White
Supplier:  Market Lab ML5367WH
Encompass_Preferred
The ML5367-WH screw caps from MarketLab are white polypropylene caps designed to fit graduated tubes. They have a leakproof design with an inner cone seal and outer o-ring seal to ensure a tight fit. The caps have a diameter of 16mm and feature a knurled grip pattern for easy opening and closing. They are autoclavable up to 121°C and can withstand temperatures from -20°C to 100°C. These screw caps are sold in packs of 100.
Catalog No. NC2398552
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.