Learn More
Cytiva SCREW LID GL45 KIT, AKTA
Supplier:  Cytiva 11000410
Encompass
Bottle top for buffer bottles used with KTA systems. Lid is polypropylene sized for GL45 glass bottles with membrane (EPDM) that allows tubing to be passed through. Not for use with organic solvents. One per pack.0
Catalog No. 50-105-5643