Learn More
United Scientific Supplies Screw Post Knife Switch
SDPSCI_ED
Catalog No. S27818 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Plastic Base with Thumbscrew Connecting Posts
Catalog No. Includes
S27818 Plastic Base with Thumbscrew Connecting Posts
1 options
Catalog No. S27818 Supplier United Scientific Supplies Supplier No. KSW002
Add to Cart
Add to Cart
Suitable for experiments requiring less than 3.6 V
  • Plastic base with included thumbscrew connecting posts
Specifications
Product Type Knife Switch
For Use With (Application) Experiments That Require Less Than 3.6 V
Includes Plastic Base with Thumbscrew Connecting Posts