Learn More
Thermo Scientific™ Screw Vial Caps and Seals
Description
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Specifications
Specifications
| Description | Black plastic cap |
| Color | Black |
| Material | Plastic |
| For Use With (Equipment) | Thermo Scientific Spectra System AS3500/AS3000/AS1000/Surveyor/Accela LC, Agilent 1090, PE Autosystem |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.