Learn More
Globe Scientific Screwcap Microcentrifuge Tubes
Catalog No. 21402940
Change view
Click to view available options
Bottom Shape:
Conical
Conical (Self-standing)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape
21-402-940 Conical
21-402-941 Conical (Self-standing)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-940 Supplier Globe Scientific Supplier No. 111710
Only null left
Add to Cart
Add to Cart
The non-sterile tubes and caps are sold separately so that you have a choice of cap styles (with o-ring or without)
  • These disposable screwcap microcentrifuge tubes are available sterile or non-sterile
  • The sterile tubes include a cap with a clear (natural) nitrile o-ring
  • All are DNase, RNase and Pyrogen free
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
Sterility Sterile
Graduated Yes
Closure Type Screw Cap with O-ring (Attached)
Bottom Shape Conical
Shape Cylindrical
Product Type Microcentrifuge Tube