Learn More
Globe Scientific Screwcap without O-Ring for Sample Tubes
Catalog No. 21403111
Change view
Click to view available options
Color:
Blue
Gray
Green
Orange
Purple
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-403-111 Gray
21-403-112 Blue
21-403-113 Green
21-403-114 Purple
21-403-115 Orange
21-403-116 Red
21-403-117 White
21-403-118 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 21-403-111 Supplier Globe Scientific Supplier No. 6136A
Only null left
Add to Cart
Add to Cart
Designed for the storage and transportation of biological material, manufactured from non-toxic polypropylene and offer strength and clarity
  • The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
  • The caps are available with or with out a silicone o-ring and are available in a variety of colors. Screw caps sold separately
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Specifications
Product Type Screw Cap
Autoclavable Autoclavable
Material Polypropylene
Color Gray
Sterility Non-sterile