Learn More
Description
- For use with the Globe Scientific sample tubes (21-200-102)
- Polypropylene construction provides the same coefficient of expansion as the tubes they fit
- Polypropylene inner lip ensures a leakproof seal even at changing temperatures
- Features a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn
Specifications
Specifications
| Material | Polypropylene |
| Color | Natural |
| Quantity | 1000/Pk. |
| For Use With (Equipment) | Globe Scientific sample tubes with external threads (21-200-102) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.