Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Scriptaid, HDAC Inhibitor, 326.35 g/mol, >98% Purity, 10 mg

Catalog No. 502707173
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7173 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7173 Supplier Abcam Supplier No. AB12088310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Novel, less toxic HDAC inhibitor that induces cell cycle arrest with >98% purity.

  • Molecular Weight: 326.35 g/mol
  • Purity: >98%
  • Description: Novel HDAC inhibitor that causes a >100-fold increase in histone deacetylation
  • Key Features: Induces cell cycle arrest in vitro and in vivo; exhibits lower toxicity than trichostatin A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Scriptaid
Content And Storage Store at -20°C.
Shipping Condition Ambient
Target Molecule HDAC
Quantity 10 mg
For Use With (Application) HDAC inhibition, histone deacetylation research, cell cycle arrest
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.