Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Scriptaid, HDAC Inhibitor, Purity >98%, 25 mg
Description
- Molecular Weight: 326.35 g/mol
- Purity: >98%
- Function: Novel HDAC inhibitor; causes >100-fold increase in histone deacetylation; induces cell cycle arrest in vitro and in vivo
- Comparison: Less toxic than trichostatin A (ab120850)
- CAS Number: 287383-59-9
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | HDAC |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Format | Vial |
| Quantity | 25 mg |
| For Use With (Application) | HDAC inhibition, cell cycle arrest |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?