Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568498 Supplier Invitrogen™ Supplier No. PA568498
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 93%; Human: 100%; Mouse: 93%; Rat: 93%.

This gene is a member of the Snail family of C2H2-type zinc finger transcription factors. It codes for a neural-specific transcriptional repressor that binds to E-box motifs. The protein may promote neural differention and may be involved in cancers with neuroendocrine features.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Scrt1
Gene Accession No. 0
Gene Alias mScrt; scratch 1; scratch family transcriptional repressor 1; scratch family zinc finger 1; scratch homolog 1 zinc finger protein; scratch homolog 1, zinc finger protein; scratch homolog 1, zinc finger protein (Drosophila); SCRT; Scrt1; Transcriptional repressor scratch 1; ZNF898
Gene Symbols Scrt1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat SCRT1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366951
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.