Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCUBE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520991 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520991 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520991 Supplier Invitrogen™ Supplier No. PA520991
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Daudi cell lysate. PA5-20991 can be used with blocking peptide PEP-1105.

SCUBE2 is a member of a family of secreted glycoproteins that contain N-terminal EGF-like repeats and C-terminal cysteine-rich motifs and CUB domain and are expressed in embryonic neuroectoderm and vascular epithelium. SCUBE2 is a close homolog to SCUBE1 and can form a heterodimeric complex with SCUBE1. SCUBE2 is thought to participate in the Hedgehog signaling pathway by specifically interacting with Sonic Hedgehog (SHH) and the receptor PTCH1, enhancing the SHH signaling activity in the raft microdomains of the plasma membrane, and may thus play important roles in organ development and tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCUBE2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SCUBE2
Gene Accession No. Q9NQ36
Gene Alias CEGB1; CEGF1; CEGP1; Protein CEGP1; RGD1563998; Scube/You; SCUBE2; Scube2-ps1; signal peptide; Signal peptide, CUB and EGF-like domain-containing protein 2; signal peptide, CUB domain and EGF like domain containing 2; signal peptide, CUB domain, EGF-like 2; signal peptide, CUB domain, EGF-like 2, pseudogene 1
Gene Symbols SCUBE2
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human SCUBE2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499241, 57758
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.