Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scythe/Bat3 Polyclonal Antibody, Rockland™
SDP

Catalog No. p-7191131
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0115 25 μL
50-199-0114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0115 Supplier Rockland Immunochemicals Supplier No. 100401B63S
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. The antibody detects endogenous Scythe/BAT3 in cell lysates.

This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Scythe/Bat3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 80 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Bag6
Gene Accession No. P46379, Q6MG49, Q9Z1R2
Gene Alias 2410045D21Rik; AA408914; BAG 6; BAG family molecular chaperone regulator 6; BAG6; BAG-6; BAT 3; Bat3; BCL2 associated athanogene 6; BCL2-associated athanogene 6; D17H6S52E; D6S52E; DAQB-195H10.3; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; Protein G3; Protein Scythe; Scythe
Gene Symbols Bag6
Host Species Rabbit
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal portion of mouse Scythe/BAT3.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224727, 7917, 94342
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.