Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDA 3-A Ethanol, 190 Proof (Denatured), Decon™ Labs
GSA_VAGREENER_CHOICE

Catalog No. 04355151 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
4 x 1 gal.
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
04-355-151 4 x 1 gal. Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-355-151 Supplier Decon Laboratories, Inc. Supplier No. 2901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product 04-355-151: May require a federal tax exempt certificate.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Clear
Quantity 4 x 1 gal.
Packaging Plastic Bottle
Boiling Point 78.3°C
Percent Purity 0%
Grade Laboratory
Chemical Name or Material Ethanol, 190 Proof
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.