Learn More
Selleck Chemical LLC SDMA
SDP
Supplier:  Selleck Chemical LLC S58385MG
Encompass_Preferred
SMDA (symmetric dimethylarginine) is a methylated form of arginine found within all nucleated cells that is released into circulation after proteolysis then excreted through the kidneys and correlates well with GFR (glomerular filtration rate) in people dogs and cats
Catalog No. 50-000-03527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.