Learn More
Biotang Inc SDS(20%) 250ML, BM-230. 20% SDS. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped at room temperature. Store at room temperature.
Supplier:  Biotang Inc BTBB079
Encompass_Preferred
SDS(20%) 250ML, BM-230. 20% SDS. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-751-3793
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.