Learn More
ZING™ Enterprises SDS Binder
Each binder contains a heavy-duty ring assembly, available in 3 sizes.
Supplier: ZING™ Enterprises 6034
Description
ZING SDS Binder prominently stores important safety and training documents to comply with the new GHS/Hazard Communication requirements. Binder is constructed from high quality 100% post-consumer poly materials that reinforce sustainability initiatives as well as compliance. Binder is highly visible with its bright red/white colors and is extremely durable, chemical resistant and easy to clean.
Different sizes include – 1.5 in. (up to 300 sheets), 2.5 in. (up to 500 sheets), and 3 in. (up to 600 sheets). Binders are available in English and bilingual English/Spanish versions.
Specifications
Floor standing | |
2.5 in. | |
12 in. | |
Plastic | |
English, Spanish |
Hazard Communication | |
2.5 in. | |
1 | |
Red, White | |
SDS Management |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.