Learn More
Medex Supply SE-1200 EXPR BASIC EKG/ECG
SDP
Supplier:  Medex Supply EDNSE1200EXP.BASIC
Encompass_Preferred
Excellent cost performance with built-in high-resolution thermal printer. You can record 12 ECG traces on A4 size and US Letter-size paper.Features:8.4″ LCD screenAlphanumeric keyboardOne-touch operationEnhanced weak signal detectionComprehensive filters & anti-noise technologyReal-time ECG waveforms freezing and reviewAdvanced SEMIP algorithm for automatic measurements and interpretationBuilt-in high resolution thermal printer with external USB printer (optional) supportExtended printingLarge internal memory which can be enlarged by USB flash diskBuilt-in data management with reports reviewLAN/Wi-Fi(optional) connectionsBi-directional communication with SE-1515 Data Management SystemMulti-format reports export (optional)Barcode scanner support (optional)On-board PDF creator making file transfer simple and seamless2 Year warranty on the SE-1200 Express Basic ECG Machine main unit 1 Year warranty on the accessories o
Catalog No. NC2383423
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.