Learn More
Agilent Technologies™ Seahorse XFp Carrier Tray
Catalog No. 103057100
Change view
Click to view available options
For Use With (Equipment):
Seahorse XF HS Mini Analyzer, Seahorse XFp Analyzer
Catalog No. For Use With (Equipment)
10-305-7100 Seahorse XF HS Mini Analyzer, Seahorse XFp Analyzer
1 options
Catalog No. 10-305-7100 Supplier Agilent Technologies™ Supplier No. 103057100
Add to Cart
Add to Cart
Seahorse XFp Carrier Tray includes four reusable Seahorse XFp Carrier Trays and conforms to ANSI/SLAS Microplate Standards for footprint dimensions and well positions.
  • Seahorse XFp Carrier Tray is used to optimize suspension cell workflows on Seahorse XF HS Mini/XFp analyzers
  • Also used to position 8-well miniplates for imaging instrumentation and plate readers
Specifications
Type Stage Plate Adapter
For Use With (Equipment) Seahorse XF HS Mini Analyzer, Seahorse XFp Analyzer