Learn More
Research Products International Corp Seal Plate ColorTab Film, Non-Sterile, Assorted Colors, 100/pk
SDP
Supplier:  Research Products International Corp 202527
Encompass_Preferred
These films feature a color dot and white writing area on each end tab for convenient plate identification. End tab can be left intact even if center of film is removed from plate. Second end tab can be marked and separated for safe keeping. Clear adhesive film prevents sample evaporation. SealPlate ColorTab is a Trademark of Excel Scientific, Inc.
Films are supplied in assorted end tab colors that include: green, orange, red, lavender, and blue.
Catalog No. 50-197-8043
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.