Learn More
BRANDTECH™ BRAND™ Aluminum Sealing Film for Cold Storage and Automation
Shop All BRANDTECH Scientific ProductsDescription
- Made of pierceable aluminum
- Tight seal for diminished evaporation and sample protection
- Trimmed corners eliminates plate overhang and prevents interference with robotic equipment
- Adhesive-free area to ensure that tasks are contamination free
- DNase-, RNase-, and DNA-free
Specifications
Specifications
| Color | Foil |
| Description | RNase / DNase free |
| Material | Aluminum |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. per Pack | 100 Sheets/Box |
| Evaporation Barrier | Yes |
| Pre-slit | No |
| Product Type | Sealing Film |
| Product Line | BRAND |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.