Learn More
Eppendorf™ Sealing Mat for DWP96/2000 Deepwell Plate
GREENER_CHOICE
Catalog No. 05414211 Shop All Eppendorf North America Products
Change view
Click to view available options
No. per Pack:
500/Pk., 5 Bags x 10 Pieces
Catalog No. No. per Pack
05-414-211 500/Pk., 5 Bags x 10 Pieces
1 options
This item is not returnable. View return policy
Catalog No. 05-414-211 Supplier Eppendorf™ Supplier No. 0030127960
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-performance sealing option provides a secure and reliable closure suitable for incubation, processing, storage, or sample archiving.
  • Storage film and foil-glued seals for simple and dependable sealing during sample storage
  • Heat sealing rilm and foil for continuous locking with the best evaporation protection
  • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
  • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf plate lid: stable, flexible protection of samples during short-term storage
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description PCR clean
No. per Pack 500/Pk., 5 Bags x 10 Pieces
Plate Compatibility 96/2000 Deep Well Plate