Learn More
Microsolv Technology Corporation Sealing Mat, PTFE, for 96 Well Plates with 1ml Glass Inserts, 1 EA.
Supplier:  Microsolv Technology Corporation 9509647
Encompass_Preferred
This Sealing Mat has a PTFE coating that faces the plate and seals 96 Glass Inserts in a 96 Well Plate configuration. The Mat is solid for tight seals and the bottoms are wells for better sealing
Catalog No. 50-227-0029