Learn More
Alkali Scientific ASI™ 96 Well Square Silicone Sealing Mat, Pre-slit, Piercable, 10/pk, 50/case
SDP
Supplier:  Alkali Scientific MPD54S
Encompass_Preferred
ASI™ 96 Well Silicone Sealing Mat, 50/case are designed for 96-well storage plates and are available in piercable format. The square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.
Catalog No. 50-238-9136
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.