Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ 07-K200 Seamless Knit Lightweight Aramid Fiber Glove

Catalog No. 191201668E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-120-1668E 2X-Large Black
19-120-1668AA X-Small Blue
19-100-905 Small White
19-120-1668B Medium Red
19-120-1668C Large Yellow
19-120-1668D X-Large Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-120-1668E Supplier PIP™ Supplier No. 07K200/XXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit lightweight aramid fiber gloves provide cut protection, heat resistance and is inherently flame resistant. Primiarily used in the automotive industry, light heat applications, glass operations and metal stamping.

  • Gloves made of aramid fiber are cut-resistant, heat-resistant and inherently flame resistant
  • Seamless knit construction protects hands without sacrificing comfort or dexterity
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Reversible design allows replacing one glove at a time
  • Can be used as gloves or glove liners
  • Color coded cuff hems for easy size identification
  • Custom styles and modifications available

Specifications

Certifications/Compliance ASTM F2992-15 ANSI Cut Level A2 (Adopted by ANSI/ISEA 02/16)/ ASTM F1790-05 ANSI Cut Level 2
Construction Seamless Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Automotive, light heat applications, glass operations, metal stamping
Glove Cuff Color Black
Glove Cuff Style Knit
Model 07-K200/XXL
Series Kut-Gard™ 07-K200
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.