Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Seamless Knit Nylon Gloves with Polyurethane Coated Fingertips

Catalog No. 191304684D Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-4684D X-Large
19-130-4684A Small
19-130-4684B Medium
19-130-4684C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-4684D Supplier SHOWA™ Supplier No. BO600XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

General purpose gloves feature a snug fit and polyurethane coated fingertips for sensitivity and control. Permeable construction allows the hands to breathe and maintain temperature control.

  • Polyurethane fingertips provide touch sensitivity
  • Coating is permeable, allowing the hands to breathe
  • Deformation-resistant glove shape is maintained for an extended period of time
  • No fingerprints are left on clear, clean objects
  • Snug fit maximizes tactile feedback
  • Designed for precision applications

Specifications

Certifications/Compliance CE Category II, EN 388:2016+A1:2018 0131X, EN ISO 21420:2020
Certifications/Compliance (Europe) CE Category II, EN 388:2016+A1:2018 0131X, EN ISO 21420:2020
Chemotherapeutic Rated No
Construction Seamless Knit
Cuff Material Elastic
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Large
EN388 Standard 0131X
For Use With (Application) Electronic Components, Quality Control, and Integrated Circuits
Gender Unisex
Glove Cuff Color Natural
Glove Cuff Style Knit
Grip Design Smooth
Liner Material Nylon
Series BO600
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.