Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ Seamless Knit PolyKor™ Blended Gloves with Polyurethane Grip

Catalog No. 19046421 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-421 2X-Large
19-046-417 Small
19-046-418 Medium
19-046-419 Large
19-046-420 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-046-421 Supplier PIP™ Supplier No. 16645LG/XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

PolyKor™ blended shell is lightweight to provide dexterity, tactile sensitivity, and cut resistance. Polyurethane coating on the palm and fingers enhances grip capabilities in dry, or slightly oily conditions.

  • Seamless construction offers increased comfort and breathability
  • Polyurethane coatings take on the surface properties of the glove liner providing a tactile grip in dry and slightly oily conditions
  • Ideal for applications that require dexterity for precise handling
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Hi-vis coloring makes this product very easy to see
  • Color coded hems for easy size identification
  • Machine washable gloves are resistant to chemicals, water, and ultraviolet light

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material Polyurethane
Color Hi-vis Green, Gray
Cut Resistance Rating ANSI A5
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material PolyKor Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Electronics, Glass Cutting, Material Handling, and Parts Assembly
Glove Cuff Style Knit
Hem Color Blue
Series 16-645LG
Yarn Gauge 13
Length (English) 10.8 in.
Length (Metric) 27.5 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.