Learn More
Seattle Glove Gray Cut-Resistant Gloves, Polyurethane Palm Coated
Catalog No. 19670302
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-670-302 Large
19-670-300 Small
19-670-303 X-Large
19-670-301 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-670-302 Supplier Seattle Glove Supplier No. DYPU5L
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight gloves with polyurethane palm provide cut-resistance and secure grip in wet or oily environments.
  • ANSI A4 cut protection
  • Lightweight, seamless knit
  • Polyurethane coated palms
  • 13-gauge
  • Abrasion, cut and puncture resistance
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Material Polyurethane
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4444
Model DYPU5-L
Series DYPU5