Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sebacic Dihydrazide 96.0+%, TCI America™
SDP

Catalog No. S022425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S022425G 25 g
S0224500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S022425G Supplier TCI America Supplier No. S022425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 925-83-7
Molecular Formula C10H22N4O2
Molecular Weight (g/mol) 230.312
MDL Number MFCD00039763
InChI Key ZWLIYXJBOIDXLL-UHFFFAOYSA-N
Synonym Decanediohydrazide, Sebacohydrazide
PubChem CID 70219
IUPAC Name decanedihydrazide
SMILES C(CCCCC(=O)NN)CCCC(=O)NN

Specifications

Melting Point 186°C
Color White
Quantity 25 g
Formula Weight 230.31
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sebacic Dihydrazide
EINECSNumber (2)-0906
RTECSNumber VS1225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.