Learn More
Sigma Organic Chemistry Sebacoyl chloride | 250G | 111-19-3 | MFCD00000770 | 0.92
Supplier: Sigma Organic Chemistry 131784250G

Sebacoyl chloride, 250G
About This Item:
Synonym(s): Sebacyl chloride
Linear Formula: ClCO(CH2)8COCl
Molecular Weight: 239.14
CAS Number: 111-19-3
MDL Number: MFCD00000770
UNSPSC Code: 12162002
Purity: 0.92
Boiling Point: 168 C/12 mmHg (lit.)
Melting Point: -5--3 C (lit.)
Density: 1.121 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.468 (lit.)
Storage: room temp
EINECS Number: 203-843-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.