Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl 4-Hydroxybenzoate 98.0+%, TCI America™
SDP

Catalog No. H0496100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0496100G 100 g
H049625G 25 g
H0496500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0496100G Supplier TCI America Supplier No. H0496100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17696-61-6
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00059626
InChI Key ZUOTXZHOGPQFIU-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid sec-Butyl Ester, sec-Butylparaben
PubChem CID 86607
IUPAC Name butan-2-yl 4-hydroxybenzoate
SMILES CCC(C)OC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 60°C
Color White
Boiling Point 210°C
Quantity 100 g
Formula Weight 194.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material sec-Butyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.