Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. B355925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B355925ML 25 mL
B35595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B355925ML Supplier TCI America Supplier No. B355925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 819-97-6
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD15072150
InChI Key QJHDFBAAFGELLO-UHFFFAOYNA-N
Synonym Butyric Acid sec-Butyl Ester
PubChem CID 13174
ChEBI CHEBI:87318
IUPAC Name butan-2-yl butanoate
SMILES CCCC(=O)OC(C)CC

Specifications

Color Colorless
Boiling Point 152°C
Quantity 25 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Butyrate
EINECSNumber (2)-0779
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.