Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl Ethyl Ether 97.0+%, TCI America™
SDP

Catalog No. B239925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B239925G 25 g
B2399250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B239925G Supplier TCI America Supplier No. B239925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2679-87-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00053773
InChI Key VSCUCHUDCLERMY-UHFFFAOYNA-N
PubChem CID 17586
IUPAC Name 2-ethoxybutane
SMILES CCOC(C)CC

Specifications

Color Colorless
Boiling Point 82°C
Quantity 25 g
UN Number 1179
Formula Weight 102.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Ethyl Ether
RTECSNumber KN4730000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.