Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich sec-Butyl methyl ether

Catalog No. 4244635ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-446-35ML 5 mL
42-446-325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-446-35ML Supplier Sigma Aldrich Supplier No. 4244635ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sec-butyl methyl ether (2-methoxy butane) is an acyclic methyl ether
  • It is reported to be formed in trace amounts during the thermal decomposition of meso- and dl-2,3-dimethylsuccinyl peroxides
  • Sec-butyl methyl ether is prepared by the reaction of sodium sec-butylate and methyl iodide
  • Its boiling point has been predicted using back-propagation neural network (bp nn)

Specifications

CAS 6795-87-5
Density 0.742 g/mL (at 25°C (literature))
Boiling Point 57°C (760 mmHg)
Molecular Formula C5H12O
Refractive Index n20/D 1.372 (literature)
Linear Formula C2H5CH(CH3)OCH3
MDL Number MFCD00190205
Quantity 5 mL
Molecular Weight (g/mol) 88.15
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.