Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC187541000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187541000 100 mL AcroSeal™ Glass bottle
AC187548000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC187541000 Supplier Thermo Scientific Chemicals Supplier No. 187541000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-30-1 , 110-82-7 , 110-54-3
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009323
InChI Key VATDYQWILMGLEW-UHFFFAOYNA-N
Synonym sec-butyllithium, s-buli, s-butyllithium, s-butyl lithium, unii-5yv3gii1tb, lithium, 1-methylpropyl, 5yv3gii1tb, secbutyllithium, b-butyllithium, sec-butyllitium
PubChem CID 102446
IUPAC Name (butan-2-yl)lithium
SMILES [Li]C(C)CC

Specifications

Color Colorless
Linear Formula C2H5CH(CH3)Li
Fieser 09,87; 10,75; 12,97; 16,56
Density 0.7500g/mL
Solubility Information Solubility in water: reacts (forming lioh and butane).
Formula Weight 64.06
Percent Purity 10.5 to 12.5% (as BuLi) (Active base)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.75
Quantity 100 mL
Flash Point −17°C
Physical Form Liquid
Chemical Name or Material sec-Butyllithium, 1.3M Solution in Cyclohexane/hexane (92/8)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement May cause drowsiness or dizziness. Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure if inhaled. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Catches fire spontaneously if exposed to air. Reacts violently with water.
Precautionary Statement GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air.
EINECSNumber 209-927-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.