Learn More
Medchemexpress LLC HY-122720 5mg Medchemexpress, SEC inhibitor KL-1 CAS:900308-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1227205MG
Encompass_Preferred
Medchemexpress, HY-122720 5mg SEC inhibitor KL-1 CAS:900308-84-1 SEC inhibitor KL-1 (KL-1), a peptidomimetic lead compound, is a potent, selective super elongation complex (SEC) inhibitor and disrupts the interaction between the SEC scaffolding protein AFF4 and P-TEFb, resulting in impaired release of Pol II from promoter-proximal pause sites and a reduced average rate of processive transcription elongation. SEC inhibitor KL-1 exhibits an dose-dependent inhibitory effect on AFF4-CCNT1 interaction with a Ki of 3.48 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9638
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.