Learn More
Seca Corp 876 Digital flat scale with high capacity
Supplier: Seca Corp 8761321004
Scales intended for mobile use have to be lightweight. The compact seca 876 weighs only 9.3 lbs, nevertheless is very stable and suitable for patients weighing up to 550 lbs. It stands firmly on four leveling bases that guarantee stability, even when small children are weighed while held by an adult. The mother+child function determines weight with just the touch of a button. • Precise weighing technology for medical use • May be used standing up thanks to the foot pedals and tap-on function • Support feet for increased stability • Can weigh approx. 10,000 times with a single set of batteries • Mother+child function to weigh toddlers • Capacity 550 lbs / 250 kg • Graduation 0.2 lb < 330 lbs > 0.5 lb / 100 g < 150 kg > 200 g • Functions • Mother+child function, tap-on automatic switch-on, lbs/kg switch-over, automatic switch-off
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.