Learn More
California Pacific Lab & Consulting SECONDARY CONTAINER 2 GAL
Supplier: California Pacific Lab & Consulting SC30010
SECONDARY LIQUID WASTE CONTAINER, 15.5 L, HDPE, FOR JUSTRITE 2 GALLON POLYETHYLENE SAFETY CAN. CAN BE USED AS A SPRILL TRAY FOR THE 5 GALLON SAFEY CAN. HEIGHT: 6.5 in., BOTTOM OD: 14.5 in., TOP OD: 16 in.