Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Secretin (rat)
GREENER_CHOICE

Catalog No. 19191
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-191 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-191 Supplier Tocris Bioscience™ Supplier No. 1919/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gastrointestinal peptide

Secretin (rat) is a gastrointestinal peptide that stimulates pancreatic and biliary secretion. Also thought to play a role in the regulation of the hypothalamus-pituitary-adrenal axis.

Secretin (human) also available.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.