Learn More
Athens Research And Technology Inc SECRETORY IGA HU COLOSTRUM 1MG
Supplier: Athens Research And Technology Inc 16130907011MG
Secretory IgA, Human Colostrum: 1 MG; Frozen in 20 mM phosphate, pH 7.4, with 150 mM NaCl; MW 385,000; EC=1.26; Purity: > =95% by SDS-PAGE. Native; Purified from human colostrum in the Athens Research & Tech lab.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.