Learn More
ACROBiosystems Recombinant Protein;Human SECTM1 Protein;HEK293;1MG;SE1,SECTM1,K12,SECTM-1,K-12
Supplier:  ACROBiosystems SE1H52271MG
Encompass_Preferred
Recombinant Protein;Human SECTM1 Protein;1MG;Human SECTM1, His Tag (SE1-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 29 - Gly 145 (Accession # AAH17716).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.5 kDa. The protein migrates as 18-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8467
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.