Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SecurAlert Adhesive Back Badge

Catalog No. 19103177 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Application):
Contractor
Visitor
Product Type:
1 week expiration
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Product Type
19-103-177 Contractor 1 week expiration
19-103-175 Visitor 1 week expiration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-103-177 Supplier Brady™ Supplier No. 95675
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Better control visitor authorizations. Brady™ SecurAlert adhesive-back badges expire automatically to prevent unauthorized use.

  • Automatically expire to prevent unauthorized use
  • Once the "front" is placed on the "back" badge, the expiration process is started and cannot be reversed
  • Easy-to-see color change confirms whether or not a visitor is authorized to be on your premises
  • Self-adhesive back for easy application
  • 500 per pack

Specifications

Color Red on white
Dimensions (L x W) 3 in. x 2 in.
For Use With (Application) Contractor
Product Type 1 week expiration
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.