Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Secure Click™ Cartridges and Filters

Catalog No. 19072217 Shop All 3M Products
Change view
Click to view available options
Cartridge Type:
Multi-Gas, Vapor
OV
Organic Vapor
Organic Vapor, Acid Gas
Filter Type:
None
P
P100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Filter Type
19-072-217 OV P
19-072-218 Multi-Gas, Vapor None
19-072-223 Organic Vapor None
19-072-219 Organic Vapor, Acid Gas None
19-072-222 Multi-Gas, Vapor P100
19-072-220 Organic Vapor, Acid Gas P100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-072-217 Supplier 3M™ Supplier No. 7100153232
Add to Cart
Add to Cart

Secure Click™ cartridges and filters provide NIOSH-approved respiratory protection and utilize dual flow technology for easier breathing.

  • Secure Click™ connection style enables quick, and simple connection of cartridges, filters
  • Dual-flow provides easier breathing and greater comfort
  • NIOSH-approved protection against certain organic vapors, gases, and non-oil based particulates
  • Swept-back design enhances field of view and comfort
  • For workplace, and occupational applications only
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type OV
Certifications (NIOSH) NIOSH-approved
Connection Type Secure Click™
Color Black/Magenta
Filter Type P
Product Type Cartridge/Filter Combination
Filter Efficiency 100
For Use With (Application) Seal Coatings, Spray Insulation
For Use With (Equipment) 3M™ Secure Click™ Facepieces
Includes 30 Filter Pairs
Model D80921
Dimensions (L x W x H) 121 x 93 x 34 mm
Weight (Metric) 112 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.