Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Secure Click™ Filter Retainer

Catalog No. 19072229 Shop All 3M Products
Click to view available options
:
D7N11, and D7P71 Particulate Filters

Catalog No. 19-072-229

Supplier: 3M™ 7100184498

Only null left
Add to Cart
Add to Cart

3M™ Secure Click™ Filter Retainer for holding Secure Click™ Particulate Filters

  • Designed to hold 3M™ Secure Click™ Particulate Filters D7N11 and D7P71
  • Works with 3M™ Secure Click™ Cartridges D8000 Series
  • Use one D701 filter retainer per D8000 Series cartridge
  • Allows for replacement of filters prior to change out of cartridges for cost savings
  • Change 3M™ respirator filters in accordance with a well-documented change-out schedule or anytime the filters are dirty, damaged, or difficult to breathe through
  • For workplace and occupational use only
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Silicone
Product Type Filter Retainer
For Use With (Equipment) D7N11, and D7P71 Particulate Filters
Holds 3M™ Secure Click™ Particulate Filters D7N11 and D7P71
Quantity 100/Cs.
Dimensions (L x W x H) 18 x 9 x 1 mm
Weight (Metric) 12 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.