Learn More
3M™ Secure Click™ P95 Particulate Filters
Secure Click™ particulate filter P95 adds protection when certain airborne particulates are present in the work environment in addition to certain vapors or acid gases.
Supplier:  3M™ 7100213434
 View more versions of this product
Catalog No. 19-072-234
Only null left
Add to Cart
Description
Description
  • Compatible with 3M™ Secure Click™ D800 Series Cartridges
  • Held in place by required 3M™ Secure Click™ Filter Retainer D701
  • NIOSH-approved particulate filter for respiratory protection against certain particulates
  • Reduces costs by allowing for replacement of filters prior to change out of cartridges
  • For workplace and occupational use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NIOSH-approved
White
Grinding
200 Filters
81 x 85 x 5 mm
Secure Click™
P
3M™ Secure Click™ Half Facepiece Reusable Respirators HF-800 Series
D7P71
2 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.