Learn More
3M™ Secure Click™ Respirator Organic Vapor P100 Cartridge/Filter
The Secure Click™ Organic Vapor P100 Cartridge/Filter is NIOSH-approved to help provide respiratory protection against certain organic vapors and oil and non-oil based airborne particulate contaminants.
Supplier:  3M™ D80921
Catalog No. 17-100-832
Only null left
Add to Cart
Description
Description
  • Secure Click™ connection style enables a quick, simple connection of cartridges/filters
  • Secure Click™ cartridges/filters are dual-flow to allow easy breathing and greater comfort
  • Use with NIOSH-approved 3M™ Secure Click™ Facepieces
  • NIOSH-approved for use against certain organic vapors
  • For respiratory protection against certain oil and non-oil based particulates
  • Swept-back design for enhanced field of view and comfort
  • Applications: Seal Coatings, Spray Insulation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vapor
Bayonet
Secure Click
Cartridge
100
D8000
NIOSH Approved
Black
P
No
Secure Click Half Facepiece HF-800 Reusable Respirators
60 Cartridges/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.