Learn More
3M™ Secure Click™ Respirator P100 Multi-gas/Vapor Cartridge/Filter
The Secure Click™ P100 Multi-Gas/Vapor Cartridge/Filter is NIOSH-approved to help provide respiratory protection against certain multi-gas/vapors and oil and non-oil based airborne particulate contaminants.
Supplier:  3M™ D80926
Catalog No. 17-100-819
Only null left
Add to Cart
Description
Description
  • For use with 3M™ Secure Click™ facepieces
  • Secure Click™ cartridges/filters utilize dual flow technology for easy breathing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Combination
Bayonet
Secure Click
Cartridge/Filter Combination
100
D8000
NIOSH Approved
Black
P
No
Secure Click Half Facepiece HF-800 Reusable Respirators
60 Cartridges/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.