Learn More
3M™ Secure Click™ Respirator P95 Particulate Filter with Nuisance Level Organic Vapor/Acid Gas Relief
3M™ Secure Click™ Hard Case D9000 Series Particulate Filters are approved for use with 3M™ Secure Click™ Respirators in environments containing dust, mists, fumes, or mold.
Supplier: 3M™ D3078
Description
- Hard case recommended for harsh areas for easy decontamination and protection against sparks and high heat
- Filter case designed for certain spray applications
- Secure Click™ connection allows filters to be inserted for ease of assembly
- Solid top and channeled airflow help protect filter
- Swept-back design provides wide field of view
- Nuisance level organic vapor/acid gas relief
- Flame and water resistant
- May be used in a variety of industries such as metalworking, grinding, sanding and welding
- Common uses of D3078 particulate filters include metal fumes produced from welding, brazing, cutting and other operations involving heating of metals, and solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances
- May also be used for liquid or oil based particles from sprays that do not emit harmful vapors
Specifications
Particulate | |
Bayonet | |
Secure Click | |
Particulate Filter | |
95% | |
D3000 |
NIOSH Approved | |
White | |
P | |
No | |
Secure Click Half Facepiece HF-800 Reusable Respirators | |
100 Filters/Pk. |