Learn More
Norell Inc 5mm, 500MHz, 7" SECURE SERIES NMR TUBE, 5 Tubes per pack INCLUDES "NORLOC WITH SECURITY BAND" CAPS
SDP
Supplier:  Norell Inc S3305000501780
Encompass_Preferred
The Secure Series™ NMR tubes feature several patented design elements that add a superior level of sample containment and isolation, safeguarding the integrity of precious or critical NMR samples and assuring secure retention of the NMR tube in the spinner turbine. A patented Security Band™ that engages and locks into the NorLoc II Security Cap™, securely joining the cap to the NMR tube. A patented proprietary surface treatment with a unique textured surface ensures precise and positive retention in Bruker, Agilent/Varian, and Jeol spinner turbines. Because of the low coefficient of thermal expansion, Secure 33 Series™ NMR tubes show a high degree of thermal shock resistance, a necessary attribute to prevent breakage when large temperature variations are expected in variable temperature studies, degassing samples through freeze-pump-thaw cycles, etc.
Catalog No. NC2832708
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.