Learn More
Norell Inc 5mm, 900MHz 7" SECURE SERIES NMR TUBE, 5 Tubes per pack INCLUDES "NORLOC WITH SECURITY BAND" CAPS
SDP
Supplier:  Norell Inc S3309000501780
Encompass_Preferred
The Secure Series™ NMR tubes feature several patented design elements that add a superior level of sample containment and isolation, safeguarding the integrity of precious or critical NMR samples and assuring secure retention of the NMR tube in the spinner turbine. A patented Security Band™ that engages and locks into the NorLoc II Security Cap™, securely joining the cap to the NMR tube. A patented proprietary surface treatment with a unique textured surface ensures precise and positive retention in Bruker, Agilent/Varian, and Jeol spinner turbines. Because of the low coefficient of thermal expansion, Secure 33 Series™ NMR tubes show a high degree of thermal shock resistance, a necessary attribute to prevent breakage when large temperature variations are expected in variable temperature studies, degassing samples through freeze-pump-thaw cycles, etc.
Catalog No. NC2832719